missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methylpiperidine, 98%, Thermo Scientific Chemicals

Catalog No. AAL1082914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1082914 25 g
AAL1082906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1082914 Supplier Thermo Scientific Chemicals Supplier No. L1082914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Optimization of Novobiocin scaffold to produce antitumor agents, Formation of carbamates and N-alkylimidazoles. Reactant for synthesis of Pyridine derivatives as CDK5 inhibitors Phenylcarbamate derivatives as ligands for nicotinic acetylcholine receptors, Amino phosphite ligands, Mexiletine enantiomers by nucleophilic aromatic substitution.

Solubility
Fully misicible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 3554-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00006495
InChI Key UKANCZCEGQDKGF-UHFFFAOYSA-N
Synonym 3-hydroxy-1-methylpiperidine, 1-methyl-3-piperidinol, n-methyl-3-piperidinol, 3-piperidinol, 1-methyl, 3-hydroxy-n-methylpiperidine, 1-methyl-3-hydroxypiperidine, n-methyl-3-hydroxypiperidine, n-methyl-3-hydroxy piperidine, 3r-1-methyl-3-piperidinol, pubchem6759
PubChem CID 98016
IUPAC Name 1-methylpiperidin-3-ol
SMILES CN1CCCC(C1)O

Specifications

Density 0.999
Boiling Point 76°C to 78°C (11 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.474
Quantity 25 g
Beilstein 103017
Solubility Information Fully misicible in water.
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-1-methylpiperidine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 222-609-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only