missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-adamantanecarboxylic Acid 97.0+%, TCI America™

Catalog No. p-7130941
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12551G 1 g
H12555G 5 g
H125525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H12551G Supplier TCI America Supplier No. H12551G

Chemical Identifiers

CAS 42711-75-1
Molecular Formula C11H15O3
Molecular Weight (g/mol) 195.24
MDL Number MFCD00193929
InChI Key CJJMAWPEZKYJAP-JZVMUCMXSA-M
Synonym 3-hydroxy-1-adamantanecarboxylic acid, 3-hydroxy-adamantane-1-carboxylic acid, 3-hydroxyadamantanecarboxylic acid, 1-carboxy-3-adamantanol, 3-hydroxy-1-adamantane carboxylic acid, 3-hydroxytricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3-carboxy-1-hydroxyadamantane, 3-hydroxy-tricyclo, acmc-1aqmu, 3-carboxy-1-adamantanol
PubChem CID 2736518
IUPAC Name (5R,7S)-3-hydroxyadamantane-1-carboxylate
SMILES OC12C[C@@H]3C[C@H](C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 203°C
Quantity 1 g
Formula Weight 196.25
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-1-adamantanecarboxylic Acid