missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-adamantaneacetic Acid 97.0+%, TCI America™

Catalog No. H14141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H14141G 1 g
H14145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H14141G Supplier TCI America Supplier No. H14141G

Chemical Identifiers

CAS 17768-36-4
Molecular Formula C12H17O3
Molecular Weight (g/mol) 209.27
MDL Number MFCD00167797
InChI Key JFMDWSCOQLUOCZ-LRSDLPTKSA-M
Synonym 2-3-hydroxyadamantan-1-yl acetic acid, 3-hydroxy-adamantan-1-yl-acetic acid, 3-hydroxy-1-adamantyl acetic acid, 3-hydroxy-1-adamantaneacetic acid, 2-3-hydroxy-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 decane-1-acetic acid, 3-hydroxy, 3-hydroxyadamantane-1-acetic acid, 2-3-hydroxyadamantanyl acetic acid, 3-hydroxytricyclo 3.3.1.1∼3,7∼ dec-1-yl acetic acid, 1-hydroxy-3-adamantaneacetic acid
PubChem CID 429307
IUPAC Name 2-[(5R,7S)-3-hydroxyadamantan-1-yl]acetate
SMILES OC12C[C@@H]3C[C@H](C1)CC(CC([O-])=O)(C3)C2

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 210.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-1-adamantaneacetic Acid