missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1,2-dimethyl-4(1H)-pyridone 98.0+%, TCI America™

Catalog No. p-7130819
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09445G 5 g
H094425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09445G Supplier TCI America Supplier No. H09445G
Only null left

Chemical Identifiers

CAS 30652-11-0
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00134497
InChI Key TZXKOCQBRNJULO-UHFFFAOYSA-N
Synonym deferiprone, ferriprox, 3-hydroxy-1,2-dimethyl-4 1h-pyridone, 3-hydroxy-1,2-dimethylpyridin-4 1h-one, 1,2-dimethyl-3-hydroxy-4-pyridone, 1,2-dimethyl-3-hydroxypyrid-4-one, 1,2-dimethyl-3-hydroxypyridine-4-one, cp20 chelating agent, unii-2bty8kh53l, ferriprox tn
PubChem CID 2972
ChEBI CHEBI:68554
IUPAC Name 3-hydroxy-1,2-dimethylpyridin-4-one
SMILES CC1=C(C(=O)C=CN1C)O

Specifications

Melting Point 260°C
Color White-Yellow
Quantity 5 g
Formula Weight 139.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-1,2-dimethyl-4(1H)-pyridone
RTECSNumber UU7785940
TSCA No