missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-1,2-dimethyl-4(1H)-pyridone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 260°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 139.15 |
| Percent Purity | ≥98.0% (T) |
| Grade | Reagent |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Hydroxy-1,2-dimethyl-4(1H)-pyridone |
Safety and Handling
| RTECSNumber | UU7785940 |
| TSCA | No |