missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one 95.0+%, TCI America™

Catalog No. p-7130702
Change view
Click to view available options
Quantity:
1 mL
5 mL
Catalog No. Quantity
H07411ML 1 mL
H07415ML 5 mL
2 options

Catalog No. H07411ML

Supplier: TCI America H07411ML

Only null left

Chemical Identifiers

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Specifications

Boiling Point 77°C
Quantity 1 mL
UN Number 1992
Formula Weight 96.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexyn-2-one