missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyn-2-one, 97%

Catalog No. p-7058300
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0531504 2 g
AAL0531509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0531504 Supplier Thermo Scientific Chemicals Supplier No. L0531504
Only null left

CAS: 1679-36-3 | C6H8O | 96.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1679-36-3
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00041627
InChI Key LTAPKZGQTMVYMX-UHFFFAOYSA-N
Synonym 3-hexyn-2-one, 2-oxo-3-hexyne, acmc-209dxd, 1-butynyl methyl ketone
PubChem CID 137151
IUPAC Name hex-3-yn-2-one
SMILES CCC#CC(=O)C

Specifications

Density 0.88
Boiling Point 67°C to 68°C (50 mmHg)
Flash Point 39°C (102°F)
Refractive Index 1.441
Quantity 2 g
UN Number UN1992
Beilstein 1699613
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 3-Hexyn-2-one
Hazard Category H226-H300-H315-H319-H335
Hazard Statement GHS H Statement
H331-H318-H226-H302-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only