missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97.0+%, TCI America™
$143.54 - $1673.92
Chemical Identifiers
| CAS | 850881-09-3 |
|---|---|
| Molecular Formula | C16H27BO2S |
| Molecular Weight (g/mol) | 294.26 |
| MDL Number | MFCD11045447 |
| InChI Key | XCXAUPBHQCCWCI-UHFFFAOYSA-N |
| Synonym | 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hexyl-2-thiopheneboronic Acid Pinacol Ester |
| PubChem CID | 16663552 |
| IUPAC Name | 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)CCCCCC |
Chemical Identifiers
| 850881-09-3 | |
| 294.26 | |
| XCXAUPBHQCCWCI-UHFFFAOYSA-N | |
| 16663552 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)CCCCCC |
| C16H27BO2S | |
| MFCD11045447 | |
| 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hexyl-2-thiopheneboronic Acid Pinacol Ester | |
| 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
| 850881-09-3 | |
| C16H27BO2S | |
| 1 g | |
| XCXAUPBHQCCWCI-UHFFFAOYSA-N | |
| 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 16663552 | |
| ≥97.0% (GC) | |
| 3-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene |
| Yellow | |
| MFCD11045447 | |
| 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hexyl-2-thiopheneboronic Acid Pinacol Ester | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)CCCCCC | |
| 294.26 | |
| 294.26 | |
| Liquid |
Safety and Handling
TSCA : No