missing translation for 'onlineSavingsMsg'
Learn More

3-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 97.0+%, TCI America™

Catalog No. H12981G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12981G 1 g
H12985G 5 g
H129825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H12981G Supplier TCI America Supplier No. H12981G
Only null left

Chemical Identifiers

CAS 850881-09-3
Molecular Formula C16H27BO2S
Molecular Weight (g/mol) 294.26
MDL Number MFCD11045447
InChI Key XCXAUPBHQCCWCI-UHFFFAOYSA-N
Synonym 2-(3-Hexyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Hexyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 16663552
IUPAC Name 2-(3-hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)CCCCCC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 294.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene