missing translation for 'onlineSavingsMsg'
Learn More

3-Hexanone 98.0+%, TCI America™

Catalog No. p-7130352
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01155ML 5 mL
H011525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01155ML Supplier TCI America Supplier No. H01155ML
Only null left

Chemical Identifiers

CAS 589-38-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009402
InChI Key PFCHFHIRKBAQGU-UHFFFAOYSA-N
Synonym 3-hexanone, ethyl propyl ketone, 3-oxohexane, aethylpropylketon, ethyl n-propyl ketone, 3-hexanone natural, aethylpropylketon german, fema no. 3290, 3-hexanone, analytical standard, pentanone, methyl
PubChem CID 11509
IUPAC Name hexan-3-one
SMILES CCCC(=O)CC

Specifications

Color Yellow
Boiling Point 124°C
Quantity 5 mL
UN Number 1224
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Hexanone
RTECSNumber MP1575000
TSCA Yes