missing translation for 'onlineSavingsMsg'
Learn More

3-Hexadecanone, 95%

Catalog No. AAL0509814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0509814 25 g
AAL0509806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0509814 Supplier Thermo Scientific Chemicals Supplier No. L0509814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18787-64-9 | C16H32O | 240.431 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18787-64-9
Molecular Formula C16H32O
Molecular Weight (g/mol) 240.431
MDL Number MFCD00048458
InChI Key LTMXHUUHBSCKEK-UHFFFAOYSA-N
Synonym 3-hexadecanone, ethyl n-tridecyl ketone, ethyl tridecyl ketone, acmc-209eqe, oo>>ue r eyie>>u 1/4 xi feminine
PubChem CID 87789
IUPAC Name hexadecan-3-one
SMILES CCCCCCCCCCCCCC(=O)CC

Specifications

Melting Point 41°C to 45°C
Boiling Point 140°C (2 mmHg)
Quantity 25 g
Beilstein 1769865
Formula Weight 240.43
Percent Purity 95%
Chemical Name or Material 3-Hexadecanone
EINECSNumber 242-572-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only