missing translation for 'onlineSavingsMsg'
Learn More

3-Heptyn-1-ol, 98%

Catalog No. AAB2088414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2088414 25 g
AAB2088406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2088414 Supplier Thermo Scientific Chemicals Supplier No. B2088414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14916-79-1 | C7H12O | 112.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14916-79-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039551
InChI Key PSWHODJVUOXHKA-UHFFFAOYSA-N
Synonym 3-heptyn-1-ol, acmc-209d1z, ho ch2 2c=c ch2 2ch3
PubChem CID 84693
IUPAC Name hept-3-yn-1-ol
SMILES CCCC#CCCO

Specifications

Density 0.882
Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.4555
Quantity 25 g
Beilstein 1737178
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 3-Heptyn-1-ol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 238-985-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only