missing translation for 'onlineSavingsMsg'
Learn More

3-Hepten-2-one, 95%

Catalog No. AAA1908609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1908609 10 g
AAA1908618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1908609 Supplier Thermo Scientific Chemicals Supplier No. A1908609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1119-44-4 | C7H12O | 112.17 g/mol

It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

Solubility
Soluble in alcohol and slightly soluble in water(3087 mg/L @ 25°C (est.))

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1119-44-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00015564
InChI Key JHHZQADGLDKIPM-AATRIKPKSA-N
Synonym 3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one
PubChem CID 5364578
SMILES CCC\C=C\C(C)=O

Specifications

Density 0.86
Boiling Point 156°C
Flash Point 49°C (120°F)
Refractive Index 1.445
Quantity 10 g
UN Number UN1224
Beilstein 1739239
Solubility Information Soluble in alcohol and slightly soluble in water(3087mg/L @ 25°C (est.))
IUPAC Name (E)-hept-3-en-2-one
Formula Weight 112.16
Percent Purity 97%
Chemical Name or Material 3-Hepten-2-one
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 214-278-8
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only