missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone 98.0+%, TCI America™

Supplier:  TCI America H003825ML

Catalog No. H003825ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Heptanone
-39°C
148°C
MFCD00009483
1224
NGAZZOYFWWSOGK-UHFFFAOYSA-N
heptan-3-one
7802
114.19
Liquid
106-35-4
Yellow
C7H14O
25 mL
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
CCCCC(=O)CC
114.19
CHEBI:50139
≥98.0% (GC)

Chemical Identifiers

106-35-4
114.19
NGAZZOYFWWSOGK-UHFFFAOYSA-N
7802
heptan-3-one
C7H14O
MFCD00009483
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
CHEBI:50139
CCCCC(=O)CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0547

RTECSNumber : MJ5250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions