missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone, 98%

CAS: 106-35-4 | C7H14O | 114.19 g/mol

$195.73 - $594.20

Chemical Identifiers

CAS 106-35-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009483
InChI Key NGAZZOYFWWSOGK-UHFFFAOYSA-N
Synonym 3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
PubChem CID 7802
ChEBI CHEBI:50139
IUPAC Name heptan-3-one
SMILES CCCCC(=O)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120382500
SDS View Documents
Thermo Scientific Chemicals
120382500
250 mL Glass bottle
Each for $195.73
Only null left
 
AC120380010
SDS View Documents
Thermo Scientific Chemicals
120380010
1 L Glass bottle
Each for $594.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-35-4
114.19
NGAZZOYFWWSOGK-UHFFFAOYSA-N
7802
heptan-3-one
C7H14O
MFCD00009483
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
CHEBI:50139
CCCCC(=O)CC

Specifications

106-35-4
Yellow
146.0°C to 149.0°C
Authentic
Glass bottle
1.4075 to 1.4095
MFCD00009483
01,699
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
NGAZZOYFWWSOGK-UHFFFAOYSA-N
heptan-3-one
7802
114.19
Liquid
-39.0°C
0.8100g/mL
41°C
98%
C7H14O
CH3(CH2)3COC2H5
250 mL
0.81
Solubility in water: 3.3g/L water (20° c). Other solubilities: soluble in alcohol and ether
CCCCC(=O)CC
114.19
CHEBI:50139
98%
3-Heptanone
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 203-388-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only