missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone, 98%

CAS: 106-35-4 | C7H14O | 114.19 g/mol

Supplier:  Thermo Scientific Chemicals A16977AU

 View more versions of this product

Catalog No. AAA16977AU


Only null left
Explore available promotions

Description

Description

Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

Solubility
Soluble in water at 20°C 4.3g/L.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place. Protect against electrostatic charges.
Specifications

Specifications

3-Heptanone
-37°C
147°C to 149°C
Pungent
1.409
1000 mL
506161
Soluble in water at 20°C 4.3g/L.
CCCCC(=O)CC
114.19
CHEBI:50139
98%
106-35-4
0.818
41°C (105°F)
C7H14O
MFCD00009483
UN1224
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
NGAZZOYFWWSOGK-UHFFFAOYSA-N
heptan-3-one
7802
114.19

Chemical Identifiers

106-35-4
114.19
NGAZZOYFWWSOGK-UHFFFAOYSA-N
7802
heptan-3-one
C7H14O
MFCD00009483
3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
CHEBI:50139
CCCCC(=O)CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H319
Flammable liquid and vapour.
Harmful if inhaled.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P374-P501c

H226-H320-H332

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 203-388-1

RTECSNumber : MJ5250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only