missing translation for 'onlineSavingsMsg'
Learn More

3-Heptanone, 98%

Catalog No. AAA16977AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16977AK 250 mL
AAA16977AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA16977AK Supplier Thermo Scientific Chemicals Supplier No. A16977AK

CAS: 106-35-4 | C7H14O | 114.19 g/mol

Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a flavoring agent in melon, nut, and banana flavors. It is also applied as a perfuming agent in cosmetics industry.

Solubility
Soluble in water at 20°C 4.3g/L.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place. Protect against electrostatic charges.

Chemical Identifiers

CAS 106-35-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009483
InChI Key NGAZZOYFWWSOGK-UHFFFAOYSA-N
Synonym 3-heptanone, butyl ethyl ketone, n-butyl ethyl ketone, aethylbutylketon, ethylbutylcetone, ethyl n-butyl ketone, ethylbutylketon, etilbutilchetone, eptan-3-one, ethyl butyl ketone
PubChem CID 7802
ChEBI CHEBI:50139
IUPAC Name heptan-3-one
SMILES CCCCC(=O)CC

Specifications

Melting Point -37°C
Density 0.818
Boiling Point 147°C to 149°C
Flash Point 41°C (105°F)
Odor Pungent
Refractive Index 1.409
Quantity 250 mL
UN Number UN1224
Beilstein 506161
Solubility Information Soluble in water at 20°C 4.3g/L.
Formula Weight 114.19
Percent Purity 98%
Chemical Name or Material 3-Heptanone
Show More Show Less
Hazard Category H226-H320-H332
Hazard Statement GHS H Statement
H226-H332-H319
Flammable liquid and vapour.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P374-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 203-388-1
RTECSNumber MJ5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only