missing translation for 'onlineSavingsMsg'
Learn More

3-Glycidoxypropyltrimethoxysilane, 97%

Catalog No. ac216540025
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2.5 kg
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC216540025 2.5 kg Glass bottle
AC216541000 100 g Glass bottle
2 options

Catalog No. AC216540025

Supplier: Thermo Scientific Chemicals 216540025

Only null left

CAS: 2530-83-8 | C9H20O5Si | 236.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.34
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Density 1.0700g/mL
Boiling Point 120.0°C (2.0 mmHg)
Flash Point 122°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4280 to 1.4300
Quantity 2.5 kg
Beilstein 17, V,3, 45
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, benzene and ether
Viscosity 4 mPa.s (20°C)
Formula Weight 236.34
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Glycidoxypropyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 219-784-2

RUO – Research Use Only