Learn More
(3-Glycidoxypropyl)trimethoxysilane, 97%
CAS: 2530-83-8 | C9H20O5Si | 236.339 g/mol
Supplier: Thermo Scientific Chemicals A1843122
Description
(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.
Solubility
Miscible with alcohols, ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
| 119°C to 121°C (2mmHg) | |
| <-70°C | |
| 100 g | |
| MFCD00005144 | |
| 135°C (275°F) | |
| Miscible with alcohols,ketones and aliphatic or aromatic hydrocarbons. Immiscible with water. | |
| CO[Si](CCCOCC1CO1)(OC)OC | |
| 236.339 | |
| 236.34 | |
| Odorless | |
| Moisture sensitive |
| (3-Glycidoxypropyl)trimethoxysilane | |
| 2530-83-8 | |
| C9H20O5Si | |
| 4308125 | |
| 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
| BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
| trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane | |
| 17317 | |
| 97% | |
| 1.429 | |
| 1.072 |
Chemical Identifiers
| 2530-83-8 | |
| 236.339 | |
| BPSIOYPQMFLKFR-UHFFFAOYSA-N | |
| 17317 | |
| CO[Si](CCCOCC1CO1)(OC)OC |
| C9H20O5Si | |
| MFCD00005144 | |
| 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane | |
| trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P280-P305+P351+P338-P308+P313-P310-P501c
H318-H341
EINECSNumber : 219-784-2
RTECSNumber : VV4025000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only