missing translation for 'onlineSavingsMsg'
Learn More

(3-Glycidoxypropyl)trimethoxysilane, 97%

Catalog No. AAA1843136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1843136 500 g
AAA1843114 25 g
AAA1843122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1843136 Supplier Thermo Scientific Chemicals Supplier No. A1843136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2530-83-8 | C9H20O5Si | 236.339 g/mol

(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

Solubility
Miscible with alcohols, ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.339
MDL Number MFCD00005144
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Boiling Point 119°C to 121°C (2mmHg)
Melting Point <-70°C
Quantity 500 g
Beilstein 4308125
Flash Point 135°C (275°F)
Solubility Information Miscible with alcohols,ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.
Formula Weight 236.34
Percent Purity 97%
Odor Odorless
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 1.072
Chemical Name or Material (3-Glycidoxypropyl)trimethoxysilane
Show More Show Less
Hazard Category H318-H341
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P280-P305+P351+P338-P308+P313-P310-P501c
EINECSNumber 219-784-2
RTECSNumber VV4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only