missing translation for 'onlineSavingsMsg'
Learn More

3-Furonitrile, 98%

Catalog No. AAL1401903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1401903 1 g
AAL1401906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1401903 Supplier Thermo Scientific Chemicals Supplier No. L1401903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 30078-65-0 | C5H3NO | 93.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30078-65-0
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.085
MDL Number MFCD00067042
InChI Key ZQFYJHMUAWCEBH-UHFFFAOYSA-N
Synonym 3-furonitrile, 3-furancarbonitrile, 3-cyanofuran, 3-furancarbonitrile 9ci, 3-furonitrile 8ci, acmc-1cj8r
PubChem CID 100698
IUPAC Name furan-3-carbonitrile
SMILES C1=COC=C1C#N

Specifications

Melting Point 28°C to 30°C
Boiling Point 150°C to 151°C
Flash Point 53°C (127°F)
Refractive Index 1.479
Quantity 1 g
UN Number UN2926
Beilstein 507009
Formula Weight 93.09
Percent Purity 98%
Chemical Name or Material 3-Furonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H228-H302-H312
Toxic if inhaled.
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only