missing translation for 'onlineSavingsMsg'
Learn More

3-Furoic acid, 99%

Catalog No. AC172450050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC172450050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC172450050 Supplier Thermo Scientific Chemicals Supplier No. 172450050
Only null left

CAS: 488-93-7 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 488-93-7
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005350
InChI Key IHCCAYCGZOLTEU-UHFFFAOYSA-N
Synonym 3-furoic acid, 3-furancarboxylic acid, 3-carboxyfuran, 3-furanoic acid, unii-88a3s7rg98, 3-furoate, 3-furans, 3-furanoate, pubchem6945, acmc-209kee
PubChem CID 10268
ChEBI CHEBI:30846
IUPAC Name furan-3-carboxylic acid
SMILES OC(=O)C1=COC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121°C to 122°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18, I, 439
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 112.08
Percent Purity 99%
Chemical Name or Material 3-Furoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
EINECSNumber 207-689-9
TSCA TSCA

RUO – Research Use Only