missing translation for 'onlineSavingsMsg'
Learn More

3-Furanmethanol 98.0+%, TCI America™

Catalog No. F036010G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F036010G 10 g
F036025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F036010G Supplier TCI America Supplier No. F036010G

Chemical Identifiers

CAS 4412-91-3
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005352
InChI Key STJIISDMSMJQQK-UHFFFAOYSA-N
Synonym 3-furanmethanol, furan-3-methanol, 3-furylmethanol, 3-hydroxymethyl furan, 3-furfuryl alcohol, 3-hydroxymethylfuran, 3-furyl alcohol, furan-3-yl-methanol, furan-3-yl methanol, 3-hydroxymethyl-furan
PubChem CID 20449
IUPAC Name (furan-3-yl)methanol
SMILES OCC1=COC=C1

Specifications

Color Yellow
Boiling Point 100°C
Quantity 10 g
UN Number 1987
Formula Weight 98.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Furanmethanol