missing translation for 'onlineSavingsMsg'
Learn More

3-Furaldehyde, 98%, stabilized

Catalog No. AC393260250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393260250 25 g Glass bottle
AC393260010 1 g Glass bottle
AC393260050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC393260250 Supplier Thermo Scientific Chemicals Supplier No. 393260250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 498-60-2 | C5H4O2 | 96.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 498-60-2
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.08
InChI Key AZVSIHIBYRHSLB-UHFFFAOYSA-N
Synonym 3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
PubChem CID 10351
ChEBI CHEBI:87609
IUPAC Name furan-3-carbaldehyde
SMILES C1=COC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1500g/mL
Boiling Point 145.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4950 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.15
Formula Weight 96.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Furaldehyde
Show More Show Less
Hazard Category GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

RUO – Research Use Only