missing translation for 'onlineSavingsMsg'
Learn More

3-Formylsalicylic acid hydrate, 97%

Catalog No. p-3758054
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377070010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC377070010 Supplier Thermo Scientific Chemicals Supplier No. 377070010

CAS: 1266615-78-4 | C8H6O4 | 166.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1266615-78-4
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD03804454
InChI Key YOEUNKPREOJHBW-UHFFFAOYSA-N
Synonym 3-Formyl-2-hydroxybenzoic acid hydrate, 2-Hydroxyisophthalaldehyde acid hydrate
IUPAC Name 3-formyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC(C=O)=C1O

Specifications

Melting Point 249°C to 252°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 166.13
Percent Purity 97%
Chemical Name or Material 3-Formylsalicylic acid hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only