missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorosalicylaldehyde 98.0+%, TCI America™
$130.48 - $380.27
Chemical Identifiers
| CAS | 394-50-3 |
|---|---|
| Molecular Formula | C7H5FO2 |
| Molecular Weight (g/mol) | 140.11 |
| MDL Number | MFCD00003319 |
| InChI Key | NWDHTEIVMDYWQJ-UHFFFAOYSA-N |
| Synonym | 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde |
| PubChem CID | 587788 |
| IUPAC Name | 3-fluoro-2-hydroxybenzaldehyde |
| SMILES | OC1=C(C=O)C=CC=C1F |
Chemical Identifiers
| 394-50-3 | |
| 140.11 | |
| NWDHTEIVMDYWQJ-UHFFFAOYSA-N | |
| 587788 | |
| OC1=C(C=O)C=CC=C1F |
| C7H5FO2 | |
| MFCD00003319 | |
| 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde | |
| 3-fluoro-2-hydroxybenzaldehyde |
Specifications
| 394-50-3 | |
| C7H5FO2 | |
| 1 g | |
| NWDHTEIVMDYWQJ-UHFFFAOYSA-N | |
| 3-fluoro-2-hydroxybenzaldehyde | |
| 587788 | |
| ≥98.0% (GC) | |
| 3-Fluorosalicylaldehyde |
| 70°C | |
| MFCD00003319 | |
| 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde | |
| OC1=C(C=O)C=CC=C1F | |
| 140.11 | |
| 140.11 | |
| Crystalline Powder |
Safety and Handling
TSCA : No