missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylaldehyde 98.0+%, TCI America™

Catalog No. F06251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06251G 1 g
F06255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06251G Supplier TCI America Supplier No. F06251G

Chemical Identifiers

CAS 394-50-3
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00003319
InChI Key NWDHTEIVMDYWQJ-UHFFFAOYSA-N
Synonym 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
PubChem CID 587788
IUPAC Name 3-fluoro-2-hydroxybenzaldehyde
SMILES OC1=C(C=O)C=CC=C1F

Specifications

Melting Point 70°C
Quantity 1 g
Formula Weight 140.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorosalicylaldehyde