missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylaldehyde, 98%

CAS: 394-50-3 | C7H5FO2 | 140.11 g/mol

$250.67 - $955.03

Chemical Identifiers

CAS 394-50-3
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00003319
InChI Key NWDHTEIVMDYWQJ-UHFFFAOYSA-N
Synonym 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
PubChem CID 587788
IUPAC Name 3-fluoro-2-hydroxybenzaldehyde
SMILES OC1=C(C=O)C=CC=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1932403
View Documents
Thermo Scientific Chemicals
L1932403
1 g
Each for $250.67
Only null left
 
AAL1932406
View Documents
Thermo Scientific Chemicals
L1932406
5 g
Each for $955.03
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

394-50-3
140.11
NWDHTEIVMDYWQJ-UHFFFAOYSA-N
587788
OC1=C(C=O)C=CC=C1F
C7H5FO2
MFCD00003319
3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
3-fluoro-2-hydroxybenzaldehyde

Specifications

394-50-3
C7H5FO2
1 g
Air Sensitive
NWDHTEIVMDYWQJ-UHFFFAOYSA-N
3-fluoro-2-hydroxybenzaldehyde
587788
98%
67°C to 71°C
MFCD00003319
1932806
3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
OC1=C(C=O)C=CC=C1F
140.11
140.12
3-Fluorosalicylaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only