missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorosalicylaldehyde, 98%

Catalog No. AAL1932403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1932403 1 g
AAL1932406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1932403 Supplier Thermo Scientific Chemicals Supplier No. L1932403

CAS: 394-50-3 | C7H5FO2 | 140.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 394-50-3
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00003319
InChI Key NWDHTEIVMDYWQJ-UHFFFAOYSA-N
Synonym 3-fluorosalicylaldehyde, 3-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 3-fluoro-2-hydroxy, zlchem 257, pubchem4215, 3-fluor-salicylaldehyd, acmc-209j5k, ksc497m8b, 2-hydroxy-3-fluoro-benzaldehyde
PubChem CID 587788
IUPAC Name 3-fluoro-2-hydroxybenzaldehyde
SMILES OC1=C(C=O)C=CC=C1F

Specifications

Melting Point 67°C to 71°C
Quantity 1 g
Beilstein 1932806
Sensitivity Air Sensitive
Formula Weight 140.12
Percent Purity 98%
Chemical Name or Material 3-Fluorosalicylaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only