missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine, 99%

Catalog No. p-3758030
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188300010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188300010 Supplier Thermo Scientific Chemicals Supplier No. 188300010
Only null left

CAS: 372-47-4 | C5H4FN | 97.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 372-47-4
MDL Number MFCD00006374
InChI Key CELKOWQJPVJKIL-UHFFFAOYSA-N
Synonym pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
PubChem CID 67794
IUPAC Name 3-fluoropyridine
SMILES C1=CC(=CN=C1)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.1300g/mL
Boiling Point 107.0°C to 108.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H4FN
Refractive Index 1.4720 to 1.4740
Quantity 1 g
Beilstein 20, IV, 2492
Specific Gravity 1.13
Molecular Weight (g/mol) 97.09
Formula Weight 97.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Fluoropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or
EINECSNumber 206-755-4

RUO – Research Use Only