missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxylic acid, 98%

Catalog No. AAH6145803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6145803 1 g
AAH6145806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6145803 Supplier Thermo Scientific Chemicals Supplier No. H6145803
Only null left

CAS: 152126-31-3 | C6H4FNO2 | 141.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 152126-31-3
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD04114108
InChI Key IRERRSXDWUCFIY-UHFFFAOYSA-N
Synonym 3-fluoropicolinic acid, 3-fluoro-2-pyridinecarboxylic acid, 3-fluoro-2-carboxypyridine, 2-pyridinecarboxylic acid, 3-fluoro, 2-pyridinecarboxylicacid, 3-fluoro, 3-fluoro-pyridine-2-carboxylic acid, 3-fluoropyridine-2-carboxylicacid, pubchem5140, acmc-1c4oi, 3-fluoropyridine-carboxylic
PubChem CID 2762786
IUPAC Name 3-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)F

Specifications

Quantity 1 g
Formula Weight 141.1
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only