missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxaldehyde, 98%

Catalog No. AAH6190206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6190206 5 g
AAH6190203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6190206 Supplier Thermo Scientific Chemicals Supplier No. H6190206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 31224-43-8 | C6H4FNO | 125.102 g/mol

3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 31224-43-8
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.102
MDL Number MFCD07781234
InChI Key OZIMPUNGBUYCSP-UHFFFAOYSA-N
Synonym 3-fluoro-2-formylpyridine, 3-fluoropicolinaldehyde, 3-fluoro-2-pyridinecarboxaldehyde, 3-fluoropyridine-2-carboxaldehyde, 3-fluoro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 3-fluoro, 2-formyl-3-fluoropyridine, pubchem5142, acmc-209hkw, 3-fluoro-2-formyl-pyridine
PubChem CID 11344017
IUPAC Name 3-fluoropyridine-2-carbaldehyde
SMILES C1=CC(=C(N=C1)C=O)F

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 125.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Fluoropyridine-2-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only