missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoropropiophenone 97.0+%, TCI America™

Catalog No. f08735g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08735G Supplier TCI America Supplier No. F08735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 455-67-4
Color Yellow
Boiling Point 96°C
Molecular Formula C9H9FO
Quantity 5 g
MDL Number MFCD00061245
Synonym 3'-fluoropropiophenone, 3-fluoropropiophenone, 1-3-fluorophenyl propan-1-one, 1-propanone, 1-3-fluorophenyl, 3-fluoro-propiophenone, 3;-fluoropropiophenone, 3-fluoro propiophenone, ethyl 3-fluorophenyl ketone, 1-3-fluorophenyl-1-propanone, 1-3-fluorophenyl-propan-1-one
InChI Key RPMOHVRRKYJFSB-UHFFFAOYSA-N
SMILES CCC(=O)C1=CC(F)=CC=C1
IUPAC Name 1-(3-fluorophenyl)propan-1-one
Molecular Weight (g/mol) 152.17
PubChem CID 521183
Formula Weight 152.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3′-Fluoropropiophenone
Show More Show Less