missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7129629
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03495G 5 g
F034925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03495G Supplier TCI America Supplier No. F03495G
Only null left

Chemical Identifiers

CAS 652-39-1
Molecular Formula C8H3FO3
Molecular Weight (g/mol) 166.11
MDL Number MFCD00039696
InChI Key WWJAZKZLSDRAIV-UHFFFAOYSA-N
Synonym 3-fluorophthalic anhydride, 4-fluoroisobenzofuran-1,3-dione, 3-fluorophthalicanhydride, 4-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 4-fluoro, 5-fluoro-isobenzofurandione, 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluor-2-benzofuran-1,3-dion, pubchem1949, fluorophthalic anhydride
PubChem CID 69551
IUPAC Name 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES FC1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 161°C
Color White-Yellow
Boiling Point 283°C
Quantity 5 g
Formula Weight 166.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluorophthalic Anhydride