missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorocinnamic Acid 98.0+%, TCI America™
Supplier: TCI America F026425G
Specifications
| 3-Fluorocinnamic Acid | |
| 166°C | |
| C9H7FO2 | |
| 25 g | |
| RTSIUKMGSDOSTI-SNAWJCMRSA-N | |
| (E)-3-(3-fluorophenyl)prop-2-enoic acid | |
| 1551219 | |
| ≥98.0% (T) |
| 458-46-8 | |
| White-Yellow | |
| MFCD00004383 | |
| 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid | |
| C1=CC(=CC(=C1)F)C=CC(=O)O | |
| 166.151 | |
| 166.15 | |
| Crystalline Powder |
Chemical Identifiers
| 458-46-8 | |
| 166.151 | |
| RTSIUKMGSDOSTI-SNAWJCMRSA-N | |
| 1551219 | |
| C1=CC(=CC(=C1)F)C=CC(=O)O |
| C9H7FO2 | |
| MFCD00004383 | |
| 3-fluorocinnamic acid, 3-3-fluorophenyl acrylic acid, trans-3-fluorocinnamic acid, 2e-3-3-fluorophenyl prop-2-enoic acid, e-3-3-fluorophenyl acrylic acid, m-fluorocinnamic acid, trans 3-fluorocinnamic acid, 3-3-fluoro-phenyl-acrylic acid, trans-m-fluorocinnamic, e-3-3-fluorophenyl prop-2-enoic acid | |
| (E)-3-(3-fluorophenyl)prop-2-enoic acid |
Safety and Handling
TSCA : No