missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorobenzyl alcohol, 98%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Density | 1.1600g/mL |
| Boiling Point | 104°C to 105°C (22.0mmHg) |
| Flash Point | 90°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5080 to 1.511 |
| Linear Formula | FC6H4CH2OH |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 207-265-3 |
| TSCA | TSCA |
RUO – Research Use Only