missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. F07905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07905G 5 g
F079025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07905G Supplier TCI America Supplier No. F07905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 701-27-9
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042286
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
PubChem CID 2734258
IUPAC Name 3-fluorobenzene-1-sulfonyl chloride
SMILES FC1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 113°C
Quantity 5 g
UN Number 3265
Formula Weight 194.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Fluorobenzenesulfonyl Chloride