missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorobenzenesulfonyl chloride, 97%

Catalog No. p-7020991
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1314003 1 g
AAA1314006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1314003 Supplier Thermo Scientific Chemicals Supplier No. A1314003
Only null left

CAS: 701-27-9 | C6H4ClFO2S | 194.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 701-27-9
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00042286
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
Synonym 3-fluorobenzene-1-sulfonyl chloride, m-fluorobenzenesulfonyl chloride, 3-fluorobenzenesulfonylchloride, 3-fluorobenzenesulphonyl chloride, 3-fluoro-benzenesulfonyl chloride, 3-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 3-fluoro, pubchem5156, acmc-209od0, 3-fluorobenzensulfonylchloride
PubChem CID 2734258
IUPAC Name 3-fluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 7°C
Density 1.463
Boiling Point 231°C to 232°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
UN Number UN3265
Beilstein 2091631
Sensitivity Moisture sensitive
Formula Weight 194.61
Percent Purity 98%
Chemical Name or Material 3-Fluorobenzenesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only