missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoroazetidine hydrochloride, 95%

Catalog No. AAH59454MD
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59454MD 250 mg
AAH59454ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59454MD Supplier Thermo Scientific Chemicals Supplier No. H59454MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 617718-46-4 | C3H7ClFN | 111.544 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 617718-46-4
Molecular Formula C3H7ClFN
Molecular Weight (g/mol) 111.544
MDL Number MFCD05663715
InChI Key PXFUWRWCKSLCLS-UHFFFAOYSA-N
Synonym 3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt
PubChem CID 10125054
IUPAC Name 3-fluoroazetidine;hydrochloride
SMILES C1C(CN1)F.Cl

Specifications

Quantity 250 mg
Formula Weight 111.55
Percent Purity 95%
Chemical Name or Material 3-Fluoroazetidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only