missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-N-methylpyrid-2-ylmethylamine, Tech., Thermo Scientific™

Chemical Identifiers

CAS 884507-13-5
Molecular Formula C7H9FN2
Molecular Weight (g/mol) 140.16
MDL Number MFCD09025827
InChI Key CRJRGUJNAQPYJG-UHFFFAOYSA-N
Synonym 3-fluoro-n-methylpyrid-2-ylmethylamine, 3-fluoropyridin-2-yl methyl methyl amine, 1-3-fluoropyridin-2-yl-n-methylmethanamine, 2-pyridinemethanamine, 3-fluoro-n-methyl, 2-pyridinemethanamine,3-fluoro-n-methyl, 3-fluoro 2-pyridyl methyl methylamine, 3-fluoro-n-methylpyrid-2-ylmethylamine,tech., 3-fluoro-n-methylpyridin-2-ylmethylamine
PubChem CID 18525718
IUPAC Name 1-(3-fluoropyridin-2-yl)-N-methylmethanamine
SMILES CNCC1=C(F)C=CC=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC30946CB
SDS View Documents
Thermo Scientific Chemicals
CC30946CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC30946DA
SDS View Documents
Thermo Scientific Chemicals
CC30946DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

884507-13-5
140.16
CRJRGUJNAQPYJG-UHFFFAOYSA-N
18525718
CNCC1=C(F)C=CC=N1
C7H9FN2
MFCD09025827
3-fluoro-n-methylpyrid-2-ylmethylamine, 3-fluoropyridin-2-yl methyl methyl amine, 1-3-fluoropyridin-2-yl-n-methylmethanamine, 2-pyridinemethanamine, 3-fluoro-n-methyl, 2-pyridinemethanamine,3-fluoro-n-methyl, 3-fluoro 2-pyridyl methyl methylamine, 3-fluoro-n-methylpyrid-2-ylmethylamine,tech., 3-fluoro-n-methylpyridin-2-ylmethylamine
1-(3-fluoropyridin-2-yl)-N-methylmethanamine

Specifications

884507-13-5
100.0
C7H9FN2
250mg
CRJRGUJNAQPYJG-UHFFFAOYSA-N
1-(3-fluoropyridin-2-yl)-N-methylmethanamine
18525718
Tech.
3-Fluoro-N-methylpyrid-2-ylmethylamine
80.0
Amber Glass Bottle
MFCD09025827
3-fluoro-n-methylpyrid-2-ylmethylamine, 3-fluoropyridin-2-yl methyl methyl amine, 1-3-fluoropyridin-2-yl-n-methylmethanamine, 2-pyridinemethanamine, 3-fluoro-n-methyl, 2-pyridinemethanamine,3-fluoro-n-methyl, 3-fluoro 2-pyridyl methyl methylamine, 3-fluoro-n-methylpyrid-2-ylmethylamine,tech., 3-fluoro-n-methylpyridin-2-ylmethylamine
CNCC1=C(F)C=CC=N1
140.16
140.16
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents