missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-DL-phenylalanine 98.0+%, TCI America™

Catalog No. F01695G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F01695G 5 g
F01691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F01695G Supplier TCI America Supplier No. F01695G
Only null left

Chemical Identifiers

CAS 456-88-2
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.182
MDL Number MFCD00004273
InChI Key VWHRYODZTDMVSS-UHFFFAOYSA-N
Synonym 2-Amino-3-(3-fluorophenyl)propionic Acid, H-DL-Phe(3-F)-OH
PubChem CID 9976
IUPAC Name 2-amino-3-(3-fluorophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)F)CC(C(=O)O)N

Specifications

Color Red-White
Quantity 5 g
Formula Weight 183.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-DL-phenylalanine
RTECSNumber AY6300000
TSCA No