missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-DL-phenylalanine, 98%

CAS: 2629-54-1 | C9H10FNO2 | 183.18 g/mol

$79.96 - $308.93

Chemical Identifiers

CAS 2629-54-1
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00004273,MFCD00066450
InChI Key VWHRYODZTDMVSS-QMMMGPOBSA-N
Synonym 3-fluoro-dl-phenylalanine, 3-fluorophenylalanine, 2-amino-3-3-fluorophenyl propanoic acid, m-fluorophenylalanine, dl-3-fluorophenylalanine, h-dl-phe 3-f-oh, m-fluoro-dl-phenylalanine, alanine, 3-fluorophenyl, 3-fluoro-3-phenylalanine, dl-phenylalanine, 3-fluoro
PubChem CID 9976
IUPAC Name 2-amino-3-(3-fluorophenyl)propanoic acid
SMILES [NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0729703
View Documents
Thermo Scientific Chemicals
L0729703
1 g
Each for $79.96
Only null left
 
AAL0729706
View Documents
Thermo Scientific Chemicals
L0729706
5 g
Each for $308.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2629-54-1
183.18
VWHRYODZTDMVSS-QMMMGPOBSA-N
9976
[NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O
C9H10FNO2
MFCD00004273,MFCD00066450
3-fluoro-dl-phenylalanine, 3-fluorophenylalanine, 2-amino-3-3-fluorophenyl propanoic acid, m-fluorophenylalanine, dl-3-fluorophenylalanine, h-dl-phe 3-f-oh, m-fluoro-dl-phenylalanine, alanine, 3-fluorophenyl, 3-fluoro-3-phenylalanine, dl-phenylalanine, 3-fluoro
2-amino-3-(3-fluorophenyl)propanoic acid

Specifications

∽245°C (decomposition)
C9H10FNO2
2939807
VWHRYODZTDMVSS-QMMMGPOBSA-N
2-amino-3-(3-fluorophenyl)propanoic acid
183.18
183.18
3-Fluoro-DL-phenylalanine
2629-54-1
MFCD00004273,MFCD00066450
3-fluoro-dl-phenylalanine, 3-fluorophenylalanine, 2-amino-3-3-fluorophenyl propanoic acid, m-fluorophenylalanine, dl-3-fluorophenylalanine, h-dl-phe 3-f-oh, m-fluoro-dl-phenylalanine, alanine, 3-fluorophenyl, 3-fluoro-3-phenylalanine, dl-phenylalanine, 3-fluoro
[NH3+][C@@H](CC1=CC=CC(F)=C1)C([O-])=O
1 g
9976
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 220-104-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only