missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluoro-D-phenylalanine, 95%
CAS: 110117-84-5 | C9H10FNO2 | 183.18 g/mol
$122.33 - $483.70
Chemical Identifiers
| CAS | 110117-84-5 |
|---|---|
| Molecular Formula | C9H10FNO2 |
| Molecular Weight (g/mol) | 183.18 |
| MDL Number | MFCD00066449 |
| InChI Key | VWHRYODZTDMVSS-UHFFFAOYNA-N |
| Synonym | 3-fluoro-d-phenylalanine, d-3-fluorophenylalanine, h-d-phe 3-f-oh, 2r-2-amino-3-3-fluorophenyl propanoic acid, m-fluoro-d-phenylalanine, 3-fluoro-d-phe, d-3-fluorophe, r-2-amino-3-3-fluorophenyl propanoic acid, 3-fluor-d-phenylalanin, h-m-fluoro-d-phe-oh |
| PubChem CID | 716317 |
| SMILES | NC(CC1=CC(F)=CC=C1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6313103
|
Thermo Scientific Chemicals
H6313103 |
1 g |
N/A
|
|
|||||
|
AAH6313106
|
Thermo Scientific Chemicals
H6313106 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 110117-84-5 | |
| 183.18 | |
| VWHRYODZTDMVSS-UHFFFAOYNA-N | |
| 716317 |
| C9H10FNO2 | |
| MFCD00066449 | |
| 3-fluoro-d-phenylalanine, d-3-fluorophenylalanine, h-d-phe 3-f-oh, 2r-2-amino-3-3-fluorophenyl propanoic acid, m-fluoro-d-phenylalanine, 3-fluoro-d-phe, d-3-fluorophe, r-2-amino-3-3-fluorophenyl propanoic acid, 3-fluor-d-phenylalanin, h-m-fluoro-d-phe-oh | |
| NC(CC1=CC(F)=CC=C1)C(O)=O |
Specifications
| 110117-84-5 | |
| MFCD00066449 | |
| VWHRYODZTDMVSS-UHFFFAOYNA-N | |
| (2R)-2-amino-3-(3-fluorophenyl)propanoic acid | |
| 183.18 | |
| 183.18 | |
| Solid |
| C9H10FNO2 | |
| 3-fluoro-d-phenylalanine, d-3-fluorophenylalanine, h-d-phe 3-f-oh, 2r-2-amino-3-3-fluorophenyl propanoic acid, m-fluoro-d-phenylalanine, 3-fluoro-d-phe, d-3-fluorophe, r-2-amino-3-3-fluorophenyl propanoic acid, 3-fluor-d-phenylalanin, h-m-fluoro-d-phe-oh | |
| NC(CC1=CC(F)=CC=C1)C(O)=O | |
| 1 g | |
| 716317 | |
| 95% | |
| 3-Fluoro-D-phenylalanine |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only