missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-D-phenylalanine, 95%

Catalog No. AAH6313103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6313103 1 g
AAH6313106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6313103 Supplier Thermo Scientific Chemicals Supplier No. H6313103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110117-84-5 | C9H10FNO2 | 183.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110117-84-5
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00066449
InChI Key VWHRYODZTDMVSS-UHFFFAOYNA-N
Synonym 3-fluoro-d-phenylalanine, d-3-fluorophenylalanine, h-d-phe 3-f-oh, 2r-2-amino-3-3-fluorophenyl propanoic acid, m-fluoro-d-phenylalanine, 3-fluoro-d-phe, d-3-fluorophe, r-2-amino-3-3-fluorophenyl propanoic acid, 3-fluor-d-phenylalanin, h-m-fluoro-d-phe-oh
PubChem CID 716317
SMILES NC(CC1=CC(F)=CC=C1)C(O)=O

Specifications

IUPAC Name (2R)-2-amino-3-(3-fluorophenyl)propanoic acid
Quantity 1 g
Formula Weight 183.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Fluoro-D-phenylalanine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only