missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH6265203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6265203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6265203 Supplier Thermo Scientific Chemicals Supplier No. H6265203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 710348-94-0 | Molecular Formula: C12H15BFNO4 | Molecular Weight: 267.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 710348-94-0
Molecular Formula C12H15BFNO4
Molecular Weight (g/mol) 267.063
InChI Key CKBJFSWEKLWHTI-UHFFFAOYSA-N
Synonym 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3-fluoro-5-nitrophenyl-4,4,5,5-tetramethyl 1,3,2 dioxaborolane
PubChem CID 59770452
IUPAC Name 2-(3-fluoro-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)[N+](=O)[O-]

Specifications

Quantity 1 g
Formula Weight 267.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Fluoro-5-nitrobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only