missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-pyridinecarboxaldehyde, 97%

Catalog No. AC442170010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442170010 1 g
AC442170050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442170010 Supplier Thermo Scientific Chemicals Supplier No. 442170010
Only null left

CAS: 40273-47-0 | C6H4FNO | 125.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40273-47-0
Molecular Formula C6H4FNO
Molecular Weight (g/mol) 125.10
MDL Number MFCD06410686
InChI Key KLKQXQXSZCNWLZ-UHFFFAOYSA-N
Synonym 3-fluoroisonicotinaldehyde, 3-fluoropyridine-4-carboxaldehyde, 3-fluoro-4-formylpyridine, 3-fluoro-4-pyridinecarboxaldehyde, 4-pyridinecarboxaldehyde, 3-fluoro, 3-fluoro-pyridine-4-carbaldehyde, 3-fluoro-4-pyridinecarbaldehyde, brettphospd, pubchem5137, acmc-20a1qb
PubChem CID 2763003
IUPAC Name 3-fluoropyridine-4-carbaldehyde
SMILES FC1=C(C=O)C=CN=C1

Specifications

Density 1.2550g/mL
Boiling Point 70.0°C (25.0 Torr)
Flash Point 67°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.255
Formula Weight 125.1
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-pyridinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only