missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitroaniline 95.0+%, TCI America™

Catalog No. F07875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07875G 5 g
F07871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07875G Supplier TCI America Supplier No. F07875G
Only null left

Chemical Identifiers

CAS 2369-13-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD03412239
InChI Key KKQPNAPYVIIXFB-UHFFFAOYSA-N
Synonym 4-amino-2-fluoronitrobenzene, 3-fluoro-4-nitro-phenylamine, 3-fluoro-4-nitro-1-benzenamine, benzenamine, 3-fluoro-4-nitro, fluoronitroaniline2, pubchem4474, 3-fluoro-4-nitroanil, 3-fluor-4-nitroaniline, 4-nitro-3-fluoroanilin, acmc-1cirv
PubChem CID 75401
IUPAC Name 3-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1N)F)[N+](=O)[O-]

Specifications

Melting Point 161°C
Quantity 5 g
Formula Weight 156.12
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Fluoro-4-nitroaniline