missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxypyridine, 97%

Catalog No. AAH6460306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6460306 5 g
AAH6460303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6460306 Supplier Thermo Scientific Chemicals Supplier No. H6460306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1060805-03-9 | C6H6FNO | 127.118 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1060805-03-9
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD13185605
InChI Key XYTUUTJNWFECNS-UHFFFAOYSA-N
Synonym 3-fluoro-4-methoxy-pyridine, pyridine, 3-fluoro-4-methoxy, abbypharma ap-14-5665
PubChem CID 57336639
IUPAC Name 3-fluoro-4-methoxypyridine
SMILES COC1=C(C=NC=C1)F

Specifications

Quantity 5 g
Beilstein 19561531
Formula Weight 127.12
Percent Purity 98%
Chemical Name or Material 3-Fluoro-4-methoxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only