Learn More
3-Fluoro-4-methoxyaniline, 99%, Thermo Scientific™
$79.81 - $243.05
Chemical Identifiers
| CAS | 366-99-4 |
|---|---|
| Molecular Formula | C7H8FNO |
| Molecular Weight (g/mol) | 141.15 |
| MDL Number | MFCD00075040 |
| InChI Key | LJWAPDSCYTZUJU-UHFFFAOYSA-N |
| Synonym | 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine |
| PubChem CID | 581110 |
| IUPAC Name | 3-fluoro-4-methoxyaniline |
| SMILES | COC1=C(C=C(C=C1)N)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC375880010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC375880050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C7H8FNO | |
| MFCD00075040 | |
| 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine | |
| 3-fluoro-4-methoxyaniline |
Specifications
| 366-99-4 | |
| 100.0 | |
| Authentic | |
| C7H8FNO | |
| MFCD00075040 | |
| LJWAPDSCYTZUJU-UHFFFAOYSA-N | |
| 3-fluoro-4-methoxyaniline | |
| 581110 | |
| 99% |
| 98.5 | |
| 135°C (18.0mmHg) | |
| 98.5% min. (GC) | |
| FC6H3(NH2)OCH3 | |
| 3-fluoro-4-anisidine, 4-amino-2-fluoroanisole, 3-fluoro-p-anisidine, 3-fluoro-4-methoxy-phenylamine, benzenamine, 3-fluoro-4-methoxy, 3-fluoro-4-methoxy-aniline, 3-fluoro-4-methoxyphenylamine, 2-fluoro-4-aminoanisole, 3-fluoro-4-methoxy-anilin, 3-fluoro-4-methoxy-benzenamine | |
| COC1=C(C=C(C=C1)N)F | |
| 141.15 | |
| 141.15 | |
| 3-Fluoro-4-methoxyaniline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact
GHS Signal Word: Warning
RUO – Research Use Only