missing translation for 'onlineSavingsMsg'
Learn More

3'-Fluoro-4'-methoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7129640
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03665G 5 g
F036625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03665G Supplier TCI America Supplier No. F03665G
Only null left

Chemical Identifiers

CAS 455-91-4
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00026219
InChI Key LQASUDVYVOFKNK-UHFFFAOYSA-N
Synonym 3'-fluoro-4'-methoxyacetophenone, 3-fluoro-4-methoxyacetophenone, 1-3-fluoro-4-methoxyphenyl ethanone, 1-3-fluoro-4-methoxyphenyl ethan-1-one, ethanone, 1-3-fluoro-4-methoxyphenyl, 3'-fluoro-4-methoxyacetophenone, acetophenone, 3'-fluoro-4'-methoxy, 1-3-fluoro-4-methoxy-phenyl-ethanone, 1-3-fluoro-4-methoxyphenyl-1-ethanone, 1-3-fluoro-4-methoxy-phenyl ethanone
PubChem CID 96828
IUPAC Name 1-(3-fluoro-4-methoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)F

Specifications

Melting Point 92°C
Color White-Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 168.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Fluoro-4′-methoxyacetophenone
EINECSNumber (3)-4389
TSCA No